missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-(trichloromethyl)pyridine, 98%

Product Code. 15432749
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15432749 5 g 5g
15422749 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15432749 Supplier Thermo Scientific Alfa Aesar Supplier No. H66418.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1929-82-4 | C6H3Cl4N | 230.90 g/mol

2-Chloro-6-(trichloromethyl)pyridine is

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-6-(trichloromethyl)pyridine is

Solubility
Soluble in 10 mg/ml ethanol.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1929-82-4
Molecular Formula C6H3Cl4N
Molecular Weight (g/mol) 230.90
MDL Number MFCD00072500
InChI Key DCUJJWWUNKIJPH-UHFFFAOYSA-N
Synonym 2-chloro-6-trichloromethyl pyridine, nitrapyrin, n-serve, pyridine, 2-chloro-6-trichloromethyl, dowco-163, n-serve nitrogen stabilizer, caswell no. 217, nitrapyrine iso-french, unii-8pce86u01w, nitrapyrin ansi:bsi:iso
PubChem CID 16004
ChEBI CHEBI:81935
IUPAC Name 2-chloro-6-(trichloromethyl)pyridine
SMILES ClC1=CC=CC(=N1)C(Cl)(Cl)Cl

Specifications

Melting Point 60°C to 65°C
Color White
Flash Point 100°C (212°F)
Quantity 5 g
UN Number UN3077
Beilstein 1618997
Solubility Information Soluble in 10mg/ml ethanol.
Formula Weight 230.9
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Chloro-6-(trichloromethyl)pyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.