missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-nitrobenzothiazole, 95%

Product Code. 15427537
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427537 10 g 10g
15417537 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427537 Supplier Thermo Scientific Alfa Aesar Supplier No. H35525.09
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2407-11-6 | C7H3ClN2O2S | 214.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2407-11-6
MDL Number MFCD00022852

Specifications

Melting Point 185°C to 189°C
Molecular Formula C7H13N
Quantity 10 g
Synonym diallylmethylamine, methyldiallylamine, 2-propen-1-amine, n-methyl-n-2-propenyl, n-allyl-n-methylprop-2-en-1-amine, methylbis prop-2-en-1-yl amine, methyldiprop-2-enylamine, 2-propen-1-amine, n-methyl-n-2-propen-1-yl, diallyl methylamine, n-methyldiallylamin, n-methyldiallylamine
InChI Key WGESLFUSXZBFQF-UHFFFAOYSA-N
SMILES CN(CC=C)CC=C
IUPAC Name N-methyl-N-prop-2-enylprop-2-en-1-amine
Molecular Weight (g/mol) 111.19
PubChem CID 75501
Formula Weight 214.63
Percent Purity 95%
Chemical Name or Material 2-Chloro-6-nitrobenzothiazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.