missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-nitrobenzothiazole, 95%

Product Code. 15417537
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417537 1 g 1g
15427537 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417537 Supplier Thermo Scientific Alfa Aesar Supplier No. H35525.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 2407-11-6 | C7H3ClN2O2S | 214.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 2407-11-6
MDL Number MFCD00022852

Specifikationer

Melting Point 185°C to 189°C
Molecular Formula C7H3ClN2O2S
Quantity 1 g
Synonym 2-chloro-6-nitrobenzothiazole, 2-chloro-6-nitrobenzo d thiazole, 2-chloro-6-nitro-benzothiazole, benzothiazole, 2-chloro-6-nitro, 2-chloro-6-nitrobenzo thiazole, 6-nitro-2-chlorobenzothiazole, pubchem21727, 6-nitro-2-chlorbenzothiazol, 2-chloro-6-nitrobenzthiazole, 2-chloro-6-nitro-benzothiazol
InChI Key KUCSJGBXJBQHNI-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C1[N+](=O)[O-])SC(=N2)Cl
IUPAC Name 2-chloro-6-nitro-1,3-benzothiazole
Molecular Weight (g/mol) 214.62
PubChem CID 75476
Formula Weight 214.63
Percent Purity 95%
Chemical Name or Material 2-Chloro-6-nitrobenzothiazole
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.