missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylpyridine, 98%

Product Code. 11429606
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429606 25 g 25g
11409606 1 g 1g
11419606 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429606 Supplier Thermo Scientific Alfa Aesar Supplier No. B20174.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18368-63-3 | C6H6ClN | 127.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18368-63-3
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00006245
InChI Key GXZDYRYYNXYPMQ-UHFFFAOYSA-N
Synonym 6-chloro-2-picoline, 6-chloro-2-methylpyridine, pyridine, 2-chloro-6-methyl, 2-chloro-6-picoline, 2-picoline, 6-chloro, unii-39mx1lq68l, 2-chloro-6-methyl pyridine, 2-chloro-6-methyl-pyridine, 2-chloro-6-methy pyridine
PubChem CID 87601
IUPAC Name 2-chloro-6-methylpyridine
SMILES CC1=CC=CC(Cl)=N1

Specifications

Density 1.167
Boiling Point 64°C to 68°C (10 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.5265
Quantity 25 g
UN Number UN2810
Beilstein 107187
Formula Weight 127.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.