missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylphenol, 98%

Product Code. 11392236
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
4.16mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392236 5 g 4.16mL
11302246 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392236 Supplier Thermo Scientific Alfa Aesar Supplier No. L03617.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 87-64-9 | C7H7ClO | 142.582 g/mol

2-Chloro-6-methylphenol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-6-methylphenol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in water (24 mg/ml at 25°C).

Notes
Storage temperature: 2 - 4°C. Incompatible with Strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-64-9
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00020000
InChI Key YPNZJHFXFVLXSE-UHFFFAOYSA-N
Synonym 6-chloro-o-cresol, phenol, 2-chloro-6-methyl, o-cresol, 6-chloro, 2-methyl-6-chlorophenol, 2-chloro-6-methyl-phenol, 6-chloro cresol, 6-chlor-o-kresol, pubchem3669, 6-chloro-2-methylphenol, 2-chloro-6-methyl phenol
PubChem CID 6898
IUPAC Name 2-chloro-6-methylphenol
SMILES CC1=C(C(=CC=C1)Cl)O

Specifications

Density 1.201
Boiling Point 75°C to 76°C (12 mmHg)
Flash Point 72°C (161°F)
Refractive Index 1.545
Quantity 5 g
UN Number UN2669
Beilstein 774623
Solubility Information Soluble in water (24mg/ml at 25°C).
Formula Weight 142.59
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylphenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-6-methylphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use dry sand, dry chemical, or alcohol-resistant foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 6-Chloro-o-cresol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.