missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylnicotinamide, ≥95%, Thermo Scientific™

Product Code. 10089541
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10089541 10 g Amber Glass Bottle 10g
10500173 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10089541 Supplier Thermo Scientific Maybridge Supplier No. SPB04933EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 54957-84-5
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.596
MDL Number MFCD00173783
InChI Key QFRWKKWOCPTEKS-UHFFFAOYSA-N
Synonym 2-chloro-6-methylnicotinamide, 3-pyridinecarboxamide,2-chloro-6-methyl, 2-chloro-6-methyl-3-pyridinecarboxamide, 2-chloro-6-methylincotinamide, 6-methyl-2-chloronicotinamide
PubChem CID 2799581
IUPAC Name 2-chloro-6-methylpyridine-3-carboxamide
SMILES CC1=NC(=C(C=C1)C(=O)N)Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 170.6
Percent Purity 95%
Chemical Name or Material 2-Chloro-6-methylnicotinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.