missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylaniline, TRC

Product Code. 30364699
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30364699 1 g
30363243 100 g
30350016 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364699 Supplier Toronto Research Chemicals Supplier No. TRCC3690701G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-63-8
Molecular Formula C7 H8 Cl N
Molecular Weight (g/mol) 141.6
Synonym o-Toluidine, 6-chloro- (8CI), 2-Chloro-6-methylbenzenamine, (2-Chloro-6-methylphenyl)amine, 2-Amino-3-chlorotoluene, 2-Chloro-6-methylaniline, 2-Methyl-6-chloroaniline, 3-Chloro-2-aminotoluene, 6-Chloro-2-methylaniline, 6-Chloro-o-toluidine, NSC 60121
IUPAC Name 2-chloro-6-methylaniline
SMILES Cc1cccc(Cl)c1N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H8ClN/c1-5-3-2-4-6(8)7(5)9/h2-4H,9H2,1H3
Formula Weight 141.0345
Chemical Name or Material 2-Chloro-6-methylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.