missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methyl-3-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. 15289816 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15289816 5 g 5g
15279816 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15289816 Supplier Thermo Scientific Acros Supplier No. 452440050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1099597-74-6
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
InChI Key DIFCQOVMVHCCHY-UHFFFAOYSA-N
Synonym 2-chloro-6-methyl-3-trifluoromethyl pyridine, pyridine, 2-chloro-6-methyl-3-trifluoromethyl, 2-chloro-3-trifluoromethyl-6-methylpyridine, pyridine, 2-?chloro-?6-?methyl-?3-? trifluoromethyl ?
PubChem CID 37863457
IUPAC Name 2-chloro-6-methyl-3-(trifluoromethyl)pyridine
SMILES CC1=NC(=C(C=C1)C(F)(F)F)Cl

Specifications

Melting Point 30°C to 32°C
Boiling Point 84°C to 86°C (24.0 Torr)
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 195.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-methyl-3-(trifluoromethyl)pyridine
compliance-icons
Product Identifier
  • 2-Chloro-6-methyl-3-(trifluoromethyl)pyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.