missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methyl-3-pyridinecarbonitrile, TRC

Product Code. 30356591
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30356591 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30356591

Brand: Toronto Research Chemicals TRCC3697805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloro-6-methyl-3-pyridinecarbonitrile
CAS 28900-10-9
Molecular Formula C7H5ClN2
Quantity 5 g
Synonym 2-Chloro-6-methylnicotinonitrile, 2-Chloro-3-cyano-6-methylpyridine
InChI Formula InChI=1S/C7H5ClN2/c1-5-2-3-6(4-9)7(8)10-5/h2-3H,1H3
SMILES Cc1ccc(C#N)c(Cl)n1
IUPAC Name 2-chloro-6-methylpyridine-3-carbonitrile
Molecular Weight (g/mol) 152.58
Formula Weight 152.0141
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.