missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methoxyquinoline-3-carboxaldehyde, 99%

Product Code. 11461590
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461590 5 g 5g
11471590 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461590 Supplier Thermo Scientific Alfa Aesar Supplier No. H26973.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 73568-29-3 | C11H8ClNO2 | 221.64 g/mol

Mainly used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings. As raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mainly used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings. As raw material in organic synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73568-29-3
Molecular Formula C11H8ClNO2
Molecular Weight (g/mol) 221.64
MDL Number MFCD00160585
InChI Key TZQOMBXDCIPJKW-UHFFFAOYSA-N
Synonym 2-chloro-6-methoxyquinoline-3-carboxaldehyde, 2-chloro-6-methoxy-3-quinolinecarboxaldehyde, 3-quinolinecarboxaldehyde, 2-chloro-6-methoxy, 2-chloro-3-formyl-6-methoxyquinoline, 2-chloro-6-methoxy-3-quinolinecarbaldehyde, zlchem 990, pubchem5944, 2-chloro-3-formyl-6-methoxy quinoline, 2-chloro-6-methoxyquinolin-3-carboxaldehyde, 2-chloro-6-methoxy-quinoline-3-carbaldehyde
PubChem CID 689079
IUPAC Name 2-chloro-6-methoxyquinoline-3-carbaldehyde
SMILES COC1=CC2=CC(=C(N=C2C=C1)Cl)C=O

Specifications

Melting Point 146°C to 151°C
Flash Point −9°C (16°F)
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 221.64
Percent Purity 99%
Chemical Name or Material 2-Chloro-6-methoxyquinoline-3-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.