missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-methoxyisonicotinic acid, 97%, Thermo Scientific™

Product Code. 10334492
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10334492 1 g 1g
10405912 10 g 10g
10307734 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10334492 Supplier Thermo Scientific Maybridge Supplier No. RF03693DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 15855-06-8
Molecular Formula C7H6ClNO3
Molecular Weight (g/mol) 187.579
MDL Number MFCD00173928
InChI Key PJQBTHQTVJMCFX-UHFFFAOYSA-N
Synonym 2-chloro-6-methoxyisonicotinic acid, 2-chloro-6-methoxy-isonicotinic acid, 4-pyridinecarboxylic acid, 2-chloro-6-methoxy, 2-chloro-6-methoxy-4-pyridinecarboxylic acid, 2-chloro-6-methoxyisonicotinicacid, acmc-1bs54, ksc173i0h, 2-chloro-6-methoxypyridine-4-carboxylicacid
PubChem CID 2782124
IUPAC Name 2-chloro-6-methoxypyridine-4-carboxylic acid
SMILES COC1=NC(=CC(=C1)C(=O)O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 187.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-methoxyisonicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.