missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorophenylacetonitrile, 98%, Thermo Scientific™

Product Code. 11478853
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478853 25 g 25g
11488853 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478853 Supplier Thermo Scientific Alfa Aesar Supplier No. A11023.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloro-6-fluorophenylacetonitrile may be used in synthesis of aryl fluorinated building blocks.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75279-55-9
Molecular Formula C8H5ClFN
Molecular Weight (g/mol) 169.583
MDL Number MFCD00001900
InChI Key ZGSAFMIRVLOISC-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorophenylacetonitrile, 2-chloro-6-fluorobenzyl cyanide, 2-2-chloro-6-fluorophenyl acetonitrile, benzeneacetonitrile, 2-chloro-6-fluoro, 2-chloro-6-fluorophenyl acetonitrile, 2-6-chloro-2-fluorophenyl ethanenitrile, 2-chloro-6-fluorobenzeneacetonitrile, pubchem10416, acmc-209oxp, ksc494e1d
PubChem CID 123575
IUPAC Name 2-(2-chloro-6-fluorophenyl)acetonitrile
SMILES C1=CC(=C(C(=C1)Cl)CC#N)F

Specifications

Melting Point 40°C to 43°C
Boiling Point 85°C (2 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3439
Beilstein 7423296
Solubility Information Insoluble in water.
Formula Weight 169.59
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-fluorophenylacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.