missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorophenylacetic Acid, TRC

Product Code. 30351966
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30351966 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351966 Supplier Toronto Research Chemicals Supplier No. TRCC36637510G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloro-6-fluorophenylacetic Acid
CAS 37777-76-7
Molecular Formula C8 H6 Cl F O2
Quantity 10 g
InChI Formula InChI=1S/C8H6ClFO2/c9-6-2-1-3-7(10)5(6)4-8(11)12/h1-3H,4H2,(H,11,12)
SMILES OC(=O)Cc1c(F)cccc1Cl
IUPAC Name 2-(2-chloro-6-fluorophenyl)acetic acid
Molecular Weight (g/mol) 188.58
Formula Weight 188.004
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.