missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorophenylacetic acid, 98%

Product Code. 11428693
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428693 100 g 100g
11408693 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428693 Supplier Thermo Scientific Alfa Aesar Supplier No. A10460.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 37777-76-7 | C8H6ClFO2 | 188.582 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37777-76-7
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.582
MDL Number MFCD00004319
InChI Key GUAIAAXDEJZRBP-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorophenylacetic acid, 2-2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenylaceticacid, benzeneacetic acid, 2-chloro-6-fluoro, 2-chloro-6-fluoro-phenyl-acetic acid, pubchem4254, acmc-1aewm, rarechem al bo 0114
PubChem CID 123464
IUPAC Name 2-(2-chloro-6-fluorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1)Cl)CC(=O)O)F

Specifications

Melting Point 120°C to 124°C
Quantity 100 g
Beilstein 4401142
Formula Weight 188.59
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-fluorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.