Product Code. 10401061
Shop All Thermo Scientific Chemicals Products
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
10401061
5 g
Glass bottle
5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
10401061
Supplier
Thermo Scientific Acros
Supplier No.
203560050
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Specifications
CAS
37777-76-7
CAS Min %
97.5
CAS Max %
100.0
Melting Point
120°C to 124°C
Color
White
Infrared Spectrum
Authentic
Assay Percent Range
98%
Packaging
Glass bottle
Molecular Formula
C8H6ClFO2
Linear Formula
ClC6 H3 (F)CH2 CO2 H
MDL Number
MFCD00004319
Quantity
5 g
Synonym
2-chloro-6-fluorophenylacetic acid, 2-2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenylaceticacid, benzeneacetic acid, 2-chloro-6-fluoro, 2-chloro-6-fluoro-phenyl-acetic acid, pubchem4254, acmc-1aewm, rarechem al bo 0114
InChI Key
GUAIAAXDEJZRBP-UHFFFAOYSA-N
SMILES
C1=CC(=C(C(=C1)Cl)CC(=O)O)F
IUPAC Name
2-(2-chloro-6-fluorophenyl)acetic acid
Molecular Weight (g/mol)
188.582
PubChem CID
123464
Formula Weight
188.59
Percent Purity
≥97.5%
Physical Form
Fine Crystalline Powder
Chemical Name or Material
2-Chloro-6-fluorophenylacetic acid, 98%
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?