missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzyl alcohol, 97%

Product Code. 11454004
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454004 25 g 25g
11444004 5 g 5g
11464004 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454004 Supplier Thermo Scientific Alfa Aesar Supplier No. A11978.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56456-50-9 | C7H6ClFO | 160.572 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56456-50-9
Molecular Formula C7H6ClFO
Molecular Weight (g/mol) 160.572
MDL Number MFCD00004608
InChI Key ZZFCUESFXBCRSC-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorobenzyl alcohol, 2-chloro-6-fluorophenyl methanol, 2-chloro-6-fluorobenzylalcohol, 2-fluoro-6-chlorobenzyl alcohol, unii-5k0sc480hm, benzenemethanol, 2-chloro-6-fluoro, 6-fluoro-2-chlorobenzyl alcohol, 2-chloro-6-fluorobenzylic alcohol, pubchem3427, acmc-1axh3
PubChem CID 91868
IUPAC Name (2-chloro-6-fluorophenyl)methanol
SMILES C1=CC(=C(C(=C1)Cl)CO)F

Specifications

Melting Point 42°C to 44°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1938353
Formula Weight 160.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluorobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.