missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-fluorobenzoic acid, 98%, ACROS Organics™

Product Code. 10601453 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10601453 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10601453 Supplier Thermo Scientific Acros Supplier No. 176230050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 434-75-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 156°C to 160°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H4ClFO2
Linear Formula ClC6H3(F)CO2H
MDL Number MFCD00002417
Quantity 5 g
Synonym benzoic acid, 2-chloro-6-fluoro, unii-euq5bp41s0, euq5bp41s0, 2-fluoro-6-chlorobenzoic acid, gnf-pf-2650, 2-chloro-6-fluoro-benzoic acid, 2-chloro-6-fluorobenzoicacid, pubchem1372, acmc-1ctwn, ksc238i3p
InChI Key XNTIGDVFBDJLTQ-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)Cl)C(=O)O)F
IUPAC Name 2-chloro-6-fluorobenzoic acid
Molecular Weight (g/mol) 174.555
PubChem CID 67947
Formula Weight 174.56
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-fluorobenzoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.