missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzene-1-carbonyl chloride, 97%, Thermo Scientific™

Product Code. p-2698118
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10722937

Brand: Thermo Scientific Maybridge SPB06439DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 79455-63-3
Molecular Formula C7H3Cl2FO
Molecular Weight (g/mol) 192.998
InChI Key GFNAJZAKJGKJCS-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorobenzoylchloride, 2-chloro-6-fluorobenzene-1-carbonyl chloride, 2-chloro-6-fluoro-benzoyl chloride, 6-chloro-2-fluorobenzoyl chloride, benzoyl chloride, 2-chloro-6-fluoro, acmc-1behg, 2-fluoro-6-chlorobenzoyl chloride, attercop-chm at108371, 2-chloro-6-fluoro benzoyl chloride, 2-chloranyl-6-fluoranyl-benzoyl chloride
PubChem CID 2736547
IUPAC Name 2-chloro-6-fluorobenzoyl chloride
SMILES C1=CC(=C(C(=C1)Cl)C(=O)Cl)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 193
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluorobenzene-1-carbonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.