missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzene-1-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10260495
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10260495 25 g 25g
10722937 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10260495 Supplier Thermo Scientific Maybridge Supplier No. SPB06439EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 79455-63-3
Molecular Formula C7H3Cl2FO
Molecular Weight (g/mol) 192.998
InChI Key GFNAJZAKJGKJCS-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorobenzoylchloride, 2-chloro-6-fluorobenzene-1-carbonyl chloride, 2-chloro-6-fluoro-benzoyl chloride, 6-chloro-2-fluorobenzoyl chloride, benzoyl chloride, 2-chloro-6-fluoro, acmc-1behg, 2-fluoro-6-chlorobenzoyl chloride, attercop-chm at108371, 2-chloro-6-fluoro benzoyl chloride, 2-chloranyl-6-fluoranyl-benzoyl chloride
PubChem CID 2736547
IUPAC Name 2-chloro-6-fluorobenzoyl chloride
SMILES C1=CC(=C(C(=C1)Cl)C(=O)Cl)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 193
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluorobenzene-1-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.