missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzaldehyde, 97%

Product Code. 11428773
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428773 100 g 100g
11418773 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428773 Supplier Thermo Scientific Alfa Aesar Supplier No. A10513.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 387-45-1 | C7H4ClFO | 158.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 387-45-1
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
MDL Number MFCD00003306
InChI Key OACPOWYLLGHGCR-UHFFFAOYSA-N
Synonym benzaldehyde, 2-chloro-6-fluoro, 2,6-cfad, 2-chloro-6-fluoro-benzaldehyd, unii-51yj9bw8w7, 2-chloro-6-fluoro benzaldehyde, 2-chloro-6-fluoro-benzaldehyde, 2-fluoro-6-chloro benzaldehyde, 6-chloro-2-fluorobenzaldehyde, pubchem1434, acmc-209j1r
PubChem CID 67847
IUPAC Name 2-chloro-6-fluorobenzaldehyde
SMILES FC1=CC=CC(Cl)=C1C=O

Specifications

Melting Point 35°C to 39°C
Boiling Point 91°C to 93°C (10 mmHg)
Flash Point 101°C (213°F)
Quantity 100 g
Beilstein 2245530
Sensitivity Air Sensitive
Formula Weight 158.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluorobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.