missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-fluoro-3-methylbenzaldehyde, 98%, ACROS Organics™

Product Code. 10210113 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10210113 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10210113 Supplier Thermo Scientific Acros Supplier No. 376140010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 104451-99-2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 46.0°C to 50.0°C
Color White
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H6ClFO
Linear Formula F(Cl)C6H2(CH3)CHO
MDL Number MFCD01631360
Quantity 1 g
Synonym 2-chloro-6-fluoro-3-methyl-benzaldehyde, benzaldehyde,2-chloro-6-fluoro-3-methyl, acmc-20anyq, pubchem2882, 2-chloro-6-fluoro-m-tolualdehyde, 2-fluoro-5-methyl-6-chlorobenzaldehyde, benzaldehyde, 2-chloro-6-fluoro-3-methyl, 2-chloranyl-6-fluoranyl-3-methyl-benzaldehyde, 2-chloro-6-fluoro-3-methylbenzaldehyde
InChI Key CSYKEPRGLYWJCW-UHFFFAOYSA-N
SMILES CC1=C(C(=C(C=C1)F)C=O)Cl
IUPAC Name 2-chloro-6-fluoro-3-methylbenzaldehyde
Molecular Weight (g/mol) 172.583
PubChem CID 736334
Formula Weight 172.59
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 2-Chloro-6-fluoro-3-methylbenzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.