missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluoro-3-methoxybenzaldehyde, 97%, Thermo Scientific™

Product Code. 11481350
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481350 1 g 1g
11491350 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481350 Supplier Thermo Scientific Alfa Aesar Supplier No. H26403.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloro-6-fluorobenzaldehyde was used in the synthesis of novel copolymers of methyl 2-cyano-3-dihalophenyl-2-propenoates and styrene.

Solubility
Soluble in water 15 mg/L at 20°C . Also soluble in heptane, toluene, methanol, ethyl acetate, acetone.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149949-29-1
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.58
MDL Number MFCD04115941
InChI Key WRJUPJLEUPNEBO-UHFFFAOYSA-N
Synonym 2-chloro-6-fluoro-3-methoxy-benzaldehyde
PubChem CID 3750673
IUPAC Name 2-chloro-6-fluoro-3-methoxybenzaldehyde
SMILES COC1=C(Cl)C(C=O)=C(F)C=C1

Specifications

Melting Point 104°C to 107°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in water 15mg/L at 20°C . Also soluble in heptane,toluene,methanol,ethyl acetate,acetone.
Formula Weight 188.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluoro-3-methoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.