missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-amino-9-(2’,3’,5’-tri-O-acetyl-beta-D-ribofuranosyl)purine, TRC

Product Code. 30357956
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30357956 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357956 Supplier Toronto Research Chemicals Supplier No. TRCC3643751G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 79999-39-6
Molecular Formula C16 H18 Cl N5 O7
Quantity 1 g
Synonym 2-Chloro-6-Amino-9-(2',3',5'-Tri-O-Acetyl-B-D-Ribofuranosyl)Purine
InChI Formula InChI=1S/C16H18ClN5O7/c1-6(23)26-4-9-11(27-7(2)24)12(28-8(3)25)15(29-9)22-5-19-10-13(18)20-16(17)21-14(10)22/h5,9,11-12,15H,4H2,1-3H3,(H2,18,20,21)/t9-,11-,12-,15-/m1/s1
SMILES CC(=O)OC[C@H]1O[C@H]([C@H](OC(=O)C)[C@@H]1OC(=O)C)n2cnc3c(N)nc(Cl)nc23
IUPAC Name [(2R,3R,4R,5R)-3,4-diacetyloxy-5-(6-amino-2-chloropurin-9-yl)oxolan-2-yl]methyl acetate
Molecular Weight (g/mol) 427.8
Formula Weight 427.0895
Chemical Name or Material 2-Chloro-6-amino-9-(2',3',5'-tri-O-acetyl-beta-D-ribofuranosyl)purine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.