missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)pyridine, 97%

Product Code. 11348035
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348035 25 g 25g
11358035 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348035 Supplier Thermo Scientific Alfa Aesar Supplier No. L08207.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52334-81-3 | C6H3ClF3N | 181.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52334-81-3
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.54
MDL Number MFCD00042225
InChI Key JFZJMSDDOOAOIV-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl pyridine, 2,5-ctf, pyridine, 2-chloro-5-trifluoromethyl, ccris 6430, 5-trifluoromethyl-2-chloropyridine, unii-c9g66d966b, 2-chloro-5-trifluoropyridine, 2-chloro-5 trifluoromethyl pyridine
PubChem CID 92342
IUPAC Name 2-chloro-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC=C(Cl)N=C1

Specifications

Melting Point 29°C to 33°C
Density 1.417
Boiling Point 152°C to 153°C
Flash Point 62°C (143°F)
Quantity 25 g
UN Number UN2811
Beilstein 3649688
Formula Weight 181.55
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.