missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)phenylacetic acid, 97%

Product Code. 11334046
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334046 1 g 1g
11344046 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334046 Supplier Thermo Scientific Alfa Aesar Supplier No. L19153.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 22893-39-6 | C9H6ClF3O2 | 238.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22893-39-6
Molecular Formula C9H6ClF3O2
Molecular Weight (g/mol) 238.59
MDL Number MFCD01631471
InChI Key PDKWZFJSOMUXLE-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl phenylacetic acid, 2-2-chloro-5-trifluoromethyl phenyl acetic acid, 2-chloro-5-trifluoromethyl phenyl acetic acid, pubchem7160, acmc-20anz9, 2-chloro-5-trifluoromethyl-phenyl-acetic acid, benzeneacetic acid, 2-chloro-5-trifluoromethyl
PubChem CID 2778119
IUPAC Name 2-[2-chloro-5-(trifluoromethyl)phenyl]acetic acid
SMILES OC(=O)CC1=CC(=CC=C1Cl)C(F)(F)F

Specifications

Melting Point 112°C to 114°C
Quantity 1 g
Beilstein 2053702
Formula Weight 238.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)phenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.