missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-chloro-5-propylpyrimidine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 50 g |
Formula Weight | 156.62 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-5-propylpyrimidine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'