missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrophenyl isocyanate, 96%, Thermo Scientific™

Product Code. 11332837
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332837 1 g 1g
11342837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332837 Supplier Thermo Scientific Alfa Aesar Supplier No. L15994.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68622-16-2
Molecular Formula C7H3ClN2O3
Molecular Weight (g/mol) 198.562
MDL Number MFCD00037065
InChI Key DZMSHKRHIIDOIZ-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrophenyl isocyanate, 2-chloro-5-nitrophenylisocyanate, 2-chloro-5-nitrobenzenisocyanate, 1-chloranyl-2-isocyanato-4-nitro-benzene, benzene, 1-chloro-2-isocyanato-4-nitro-9ci
PubChem CID 3602243
IUPAC Name 1-chloro-2-isocyanato-4-nitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])N=C=O)Cl

Specifications

Melting Point 92°C to 95°C
Quantity 1 g
UN Number UN2811
Beilstein 6506805
Sensitivity Moisture sensitive
Formula Weight 198.57
Percent Purity 96%
Chemical Name or Material 2-Chloro-5-nitrophenyl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.