missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrophenol, 98%

Product Code. 15519583 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15519583 5 g 5g
15529583 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15519583 Supplier Thermo Scientific Acros Supplier No. 459040050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-10-3 | C6H4ClNO3 | 173.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-10-3
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.55
MDL Number MFCD01571825
InChI Key BUMGQSCPTLELLS-UHFFFAOYSA-N
Synonym phenol, 2-chloro-5-nitro, 6-chloro-3-nitrophenol, 2-chloro-5-nitro-phenol, pubchem1475, acmc-209mxh, 2-chloro-5-nitrofenol, 5-nitro-2-chlorophenol, 2-chloro-5-nitro phenol, ksc494c3h, 4-chloro-3-hydroxynitrobenzene
PubChem CID 69264
IUPAC Name 2-chloro-5-nitrophenol
SMILES OC1=CC(=CC=C1Cl)[N+]([O-])=O

Specifications

Melting Point 119.0°C to 121.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 173.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-nitrophenol
compliance-icons
Product Identifier
  • 2-Chloro-5-nitrophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.