missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-5-nitrobenzyl alcohol, 97%, ACROS Organics™

Product Code. 10504131 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
2g; Glass bottle
This item is not returnable. View return policy

Product Code. 10504131

Brand: Thermo Scientific Acros 182930020

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 80866-80-4
CAS Min % 96.0
CAS Max % 100.0
Melting Point 74°C to 79°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H6ClNO3
Linear Formula ClC6H3(NO2)CH2OH
MDL Number MFCD00007297
Quantity 2 g
Synonym 2-chloro-5-nitrophenyl methanol, 2-chloro-5-nitrobenzyl alcohol, 2-chloro-5-nitrobenzylalcohol, benzenemethanol, 2-chloro-5-nitro, acmc-1bkf2, nnlqydltfrxakd-uhfffaoysa, benzenemethanol,2-chloro-5-nitro, 2-chloro-5-nitro-phenyl-methanol, 2-chloro-5-nitrophenyl methanol #, 2-chloro-5-nitrophenyl methan-1-ol
InChI Key NNLQYDLTFRXAKD-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1[N+](=O)[O-])CO)Cl
IUPAC Name (2-chloro-5-nitrophenyl)methanol
Molecular Weight (g/mol) 187.579
PubChem CID 555722
Formula Weight 187.58
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 2-chloro-5-nitrobenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-5-nitrobenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.