missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzoic acid, 98+%

Product Code. 11444794
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444794 100 g 100g
11454794 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444794 Supplier Thermo Scientific Alfa Aesar Supplier No. A13754.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2516-96-3 | C7H4ClNO4 | 201.562 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2516-96-3
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
MDL Number MFCD00007294
InChI Key QUEKGYQTRJVEQC-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrobenzoic acid, benzoic acid, 2-chloro-5-nitro, 6-chloro-3-nitrobenzoic acid, 2-chloro-5-nitro-benzoic acid, 2-chloro-5-nitrobenzoicacid, mesalamine impurity m, pubchem13682, acmc-1cb0i, 2-chloro-5-nitrobenzic acid, 5-nitro-2-chlorobenzoic acid
PubChem CID 17287
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Cl

Specifications

Melting Point 166°C to 168°C
Flash Point >100°C (212°F)
Odor Odorless
Quantity 100 g
Beilstein 1877474
Formula Weight 201.57
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-5-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.