missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitro-N-(4-pyridyl)benzamide, Thermo Scientific™

Product Code. 15446399
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446399 50 mg 50mg
15436399 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446399 Supplier Thermo Scientific Alfa Aesar Supplier No. J64455.MF

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • A cell-permeable chloro-nitro-benzamido compound that acts as a potent, specific, irreversible, and high-affinity antagonist of PPAR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313516-66-4
Molecular Formula C12H8ClN3O3
Molecular Weight (g/mol) 277.664
MDL Number MFCD00121849
InChI Key FRPJSHKMZHWJBE-UHFFFAOYSA-N
Synonym 2-chloro-5-nitro-n-4-pyridinylbenzamide, 2-chloro-5-nitro-n-pyridin-4-yl benzamide, 2-chloro-5-nitro-n-4-pyridyl benzamide, benzamide, 2-chloro-5-nitro-n-4-pyridinyl, 2-chloro-5-nitro-n-4-pyridinyl-benzamide, d03bpg, d08gqc, 2-chloro-5-nitro-n-4-pyridinyl benzamide, benzamide,2-chloro-5-nitro-n-4-pyridinyl, n1-4-pyridyl-2-chloro-5-nitrobenzamide
PubChem CID 2777391
IUPAC Name 2-chloro-5-nitro-N-pyridin-4-ylbenzamide
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)NC2=CC=NC=C2)Cl

Specifications

Color White
Quantity 50 mg
Sensitivity Light sensitive
Solubility Information Soluble in DMSO at 10mg/ml. Insoluble in water.
Formula Weight 277.66
Physical Form Solid
Chemical Name or Material 2-Chloro-5-nitro-N-(4-pyridyl)benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.