missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylthiophene, 97%

Product Code. 11420227
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420227 5 g 5g
11430227 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420227 Supplier Thermo Scientific Alfa Aesar Supplier No. B21657.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17249-82-0 | C5H5ClS | 132.605 g/mol

It is used in the synthesis of dithienylcyclopentene optical molecular switches. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of dithienylcyclopentene optical molecular switches. It is also used as an active pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17249-82-0
Molecular Formula C5H5ClS
Molecular Weight (g/mol) 132.605
MDL Number MFCD01632145
InChI Key JSMMZMYGEVUURX-UHFFFAOYSA-N
Synonym thiophene, 2-chloro-5-methyl, pubchem5197, acmc-209e4p, 2-chloro-5-methyl-thiophene, ksc497i0h, 2-chloranyl-5-methyl-thiophene
PubChem CID 140208
IUPAC Name 2-chloro-5-methylthiophene
SMILES CC1=CC=C(S1)Cl

Specifications

Density 1.21
Boiling Point 155°C to 156°C
Flash Point 47°C (117°F)
Odor Characteristic
Refractive Index 1.5356
Quantity 5 g
UN Number UN1992
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 132.61
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-methylthiophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.