missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-5-methylphenol, 99%

Product Code. 10625422 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10625422 25 g Glass bottle 25g
10573341 100 g Glass bottle 100g
10455755 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10625422 Supplier Thermo Scientific Acros Supplier No. 170410250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 615-74-7 | C7H7ClO | 142.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-74-7
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
MDL Number MFCD00002175
InChI Key SMFHPCZZAAMJJO-UHFFFAOYSA-N
Synonym 6-chloro-m-cresol, phenol, 2-chloro-5-methyl, m-cresol, 6-chloro, 3-methyl-6-chlorophenol, 4-chloro-3-hydroxytoluene, 5-methyl-2-chlorophenol, 6-chloro-3-methylphenol, dsstox_cid_26558, dsstox_rid_81718
PubChem CID 12008
IUPAC Name 2-chloro-5-methylphenol
SMILES CC1=CC=C(Cl)C(O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 45.0°C to 48.0°C
Color Beige
Boiling Point 196.0°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(CH3)OH
Quantity 25 g
Beilstein 06, I, 187
Formula Weight 142.58
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloro-5-methylphenol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-5-methylphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.