missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-5-isocyanatopyridine, 97%, Thermo Scientific™

Product Code. 10104254
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10104254 5 g 5g
10452053 250 mg 250mg
10592973 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10104254 Supplier Thermo Scientific Maybridge Supplier No. MO07678DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 125117-96-6
Molecular Formula C6H3ClN2O
Molecular Weight (g/mol) 154.55
MDL Number MFCD07368563
InChI Key WHENXACISRCOHK-UHFFFAOYSA-N
PubChem CID 4961273
IUPAC Name 2-chloro-5-isocyanatopyridine
SMILES ClC1=NC=C(C=C1)N=C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym pyridine,2-chloro-5-isocyanato, acmc-20anz3, 6-chloropyridin-3-isocyanate, 2-chloro-5-isocyanato-pyridine, 6-chloro-3-pyridinyl isocyanate, pyridine, 2-chloro-5-isocyanato, 2-chloropyridin-5-yl isocyanate, 2-chloro-5-isocyanatopyridine
Formula Weight 154.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-isocyanatopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.