missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodobenzoic acid, 97%

Product Code. 11398976
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398976 100 g 100g
11378976 5 g 5g
11388976 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398976 Supplier Thermo Scientific Alfa Aesar Supplier No. L20161.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19094-56-5 | C7H4ClIO2 | 282.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19094-56-5
Molecular Formula C7H4ClIO2
Molecular Weight (g/mol) 282.46
MDL Number MFCD00079731
InChI Key GEBYSTBEDVQOTK-UHFFFAOYSA-N
Synonym 2-chloro-5-iodo-benzoic acid, benzoic acid, 2-chloro-5-iodo, pubchem3649, acmc-209eug, 5-iodo-2-chlorobenzoic acid, ksc174i1h, benzoicacid, 2-chloro-5-iodo, buttpark 100\01-43, 2-chloro-5-iodobenzoic acid
PubChem CID 519638
IUPAC Name 2-chloro-5-iodobenzoic acid
SMILES OC(=O)C1=C(Cl)C=CC(I)=C1

Specifications

Melting Point 156°C to 161°C
Quantity 100 g
Beilstein 2254110
Sensitivity Light sensitive
Formula Weight 282.46
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-iodobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.