missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodo-3-(trifluoromethyl)pyridine, 95%

Product Code. 15431489
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15431489 1 g 1g
15458846 250 mg 250mg
15441489 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15431489 Supplier Thermo Scientific Alfa Aesar Supplier No. H64329.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 887707-25-7 | C6H2ClF3IN | 307.44 g/mol

It is used as pharmaceutical intermediate and as an advanced chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate and as an advanced chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 887707-25-7
Molecular Formula C6H2ClF3IN
Molecular Weight (g/mol) 307.44
MDL Number MFCD12024302
InChI Key KUASPTUGCBGXAE-UHFFFAOYSA-N
Synonym 2-chloro-5-iodo-3-trifluoromethyl pyridine, 2-chloro-3-trifluoromethyl-5-iodo-pyridine, 2-chloro-3-trifluoromethyl-5-iodopyridine, 2-choro-5-iodo-3-trifluoromethylpyridine, 2-chloro-3-trifluoromethyl-5-iodo pyridine
PubChem CID 11500513
IUPAC Name 2-chloro-5-iodo-3-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC(I)=CN=C1Cl

Specifications

Melting Point 43°C
Quantity 1 g
Beilstein 10249610
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 307.44
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Chloro-5-iodo-3-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.