missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-formylbenzoic Acid, TRC

Product Code. 30357410
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357410 1 g 1g
30362669 500 mg -
30365485 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357410 Supplier Toronto Research Chemicals Supplier No. TRCC1976761G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1206625-81-1
Molecular Formula C8H5ClO3
Molecular Weight (g/mol) 184.57
IUPAC Name 2-chloro-5-formylbenzoic acid
SMILES OC(=O)c1cc(C=O)ccc1Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H5ClO3/c9-7-2-1-5(4-10)3-6(7)8(11)12/h1-4H,(H,11,12)
Formula Weight 183.9927
Chemical Name or Material 2-Chloro-5-formylbenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.