missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluoropyridine, 95%, Thermo Scientific™

Product Code. 11906451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11906451 1 g 1g
11916451 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11906451 Supplier Thermo Scientific Acros Supplier No. 442860010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 31301-51-6
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.53
MDL Number MFCD03095332
InChI Key QOGXQLSFJCIDNY-UHFFFAOYSA-N
Synonym 2-chloro-5-fluoro-pyridine, 2-chloro-5-fluoro pyridine, pyridine, 2-chloro-5-fluoro, zlchem 338, pubchem1097, 2-chlor-5-fluorpyridin, acmc-1aevz, 2-chloro-5-fluropyridine, ksc223e3t, tpc-py082
PubChem CID 2782801
IUPAC Name 2-chloro-5-fluoropyridine
SMILES FC1=CN=C(Cl)C=C1

Specifications

Melting Point 23°C to 24°C
Density 1.2970g/mL
Boiling Point 74°C (37.0 Torr)
Flash Point 42°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.297
Formula Weight 131.54
Percent Purity 95%
Chemical Name or Material 2-Chloro-5-fluoropyridine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-5-fluoropyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.