missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluorophenol, 98%, Thermo Scientific™

Catalog Number 10358660
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3827-49-4
C6H4ClFO
146.55
MFCD00042524
CMQOZIKIOASEIN-UHFFFAOYSA-N
2-chloro-5-fluoro-phenol, pubchem2146, 2-chloro-5-fluorphenol, acmc-209iz2, ksc226k0t, attercop-chm at137231
2724522
2-chloro-5-fluorophenol
OC1=CC(F)=CC=C1Cl
This item is not returnable. View return policy
3827-49-4
C6H4ClFO
146.55
MFCD00042524
CMQOZIKIOASEIN-UHFFFAOYSA-N
2-chloro-5-fluoro-phenol, pubchem2146, 2-chloro-5-fluorphenol, acmc-209iz2, ksc226k0t, attercop-chm at137231
2724522
2-chloro-5-fluorophenol
OC1=CC(F)=CC=C1Cl
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 183°C to 185°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(OH)Cl
Refractive Index 1.5240 to 1.526
Quantity 5 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in aqueous alkali,soluble in most common organic solvents
Formula Weight 146.55
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-fluorophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'