missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluoro-3-methylbenzoic acid, 98%, Thermo Scientific™

Product Code. 15419708
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1427416-28-1
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.58
MDL Number MFCD23712098
InChI Key AILHHXAQZADVIH-UHFFFAOYSA-N
Synonym 2-chloro-5-fluoro-3-methylbenzoicacid
PubChem CID 53430534
IUPAC Name 2-chloro-5-fluoro-3-methylbenzoic acid
SMILES CC1=C(Cl)C(=CC(F)=C1)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 188.58
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Chloro-5-fluoro-3-methylbenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'