missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(dimethylsulfamoyl)benzoic Acid, TRC

Product Code. 30456961
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456961 500 mg 500mg
30456895 100 mg 100mg
30454445 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456961 Supplier Toronto Research Chemicals Supplier No. TRCT776965500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37088-27-0
Molecular Formula C9H10NO4SCl
Molecular Weight (g/mol) 263.7
IUPAC Name 2-chloro-5-(dimethylsulfamoyl)benzoic acid
SMILES CN(C)S(=O)(=O)c1ccc(Cl)c(c1)C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H10ClNO4S/c1-11(2)16(14,15)6-3-4-8(10)7(5-6)9(12)13/h3-5H,1-2H3,(H,12,13)
Formula Weight 263.0019
Chemical Name or Material 2-Chloro-5-(dimethylsulfamoyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.