missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(cyclopropylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15491356
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1449132-64-2
C10H11BClNO3
239.462
MFCD22125077
ZXCSFHNBVHLHBP-UHFFFAOYSA-N
2-chloro-5-cyclopropylcarbamoyl phenylboronic acid, 2-chloro-5-cyclopropylcarbamoyl benzeneboronic acid
73995882
[2-chloro-5-(cyclopropylcarbamoyl)phenyl]boronic acid
B(C1=C(C=CC(=C1)C(=O)NC2CC2)Cl)(O)O
This item is not returnable. View return policy
1449132-64-2
C10H11BClNO3
239.462
MFCD22125077
ZXCSFHNBVHLHBP-UHFFFAOYSA-N
2-chloro-5-cyclopropylcarbamoyl phenylboronic acid, 2-chloro-5-cyclopropylcarbamoyl benzeneboronic acid
73995882
[2-chloro-5-(cyclopropylcarbamoyl)phenyl]boronic acid
B(C1=C(C=CC(=C1)C(=O)NC2CC2)Cl)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloro-5-(cyclopropylcarbamoyl)benzeneboronic acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 239.46
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(cyclopropylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'