missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(chloromethyl)pyridine, 96%

Product Code. 11394166
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394166 5 g 5g
11304176 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394166 Supplier Thermo Scientific Alfa Aesar Supplier No. L19284.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 70258-18-3 | C6H5Cl2N | 162.013 g/mol

2-Chloro-5-(chloromethyl)pyridine is used for the synthesis of various pharmaceutical compounds. It can also be used for the synthesis of new neonicotinoid compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-(chloromethyl)pyridine is used for the synthesis of various pharmaceutical compounds. It can also be used for the synthesis of new neonicotinoid compounds.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70258-18-3
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD00125366
InChI Key SKCNYHLTRZIINA-UHFFFAOYSA-N
Synonym 2-chloro-5-chloromethyl pyridine, pyridine, 2-chloro-5-chloromethyl, 2-chloro-5-chloromethyl-pyridine, pubchem9210, acmc-1bl9y, ksc377c3b, 2-chloro-5chloromethylpyridine, 2-chloro-5-chloromethypyridine, 5-chloromethyl-2-chloropyridine, 6-chloro-3-chloromethylpyridine
PubChem CID 155479
IUPAC Name 2-chloro-5-(chloromethyl)pyridine
SMILES C1=CC(=NC=C1CCl)Cl

Specifications

Melting Point 37°C to 42°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 1635690
Solubility Information Insoluble in water.
Formula Weight 162.02
Percent Purity 96%
Chemical Name or Material 2-Chloro-5-(chloromethyl)pyridine
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-5-(chloromethyl)pyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2-Chloro-5-(chloromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.