missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5,5-dimethyl-1,3-cyclohexanedione, 98%

Product Code. 15469115
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469115 250 mg 250mg
11309266 1 g 1g
15407927 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15469115 Supplier Thermo Scientific Alfa Aesar Supplier No. H51035.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7298-89-7 | C8H11ClO2 | 174.624 g/mol

2-Chloro-5,5-dimethyl-1,3-cyclohexanedione is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7298-89-7
Molecular Formula C8H11ClO2
Molecular Weight (g/mol) 174.624
MDL Number MFCD00010502
InChI Key VOBIHUAWDXUCPH-UHFFFAOYSA-N
Synonym chlorodimedone, 2-chlorodimedone, monochlorodimedone, 2-chloro-5,5-dimethyl-1,3-cyclohexanedione, 1,1-dimethyl-4-chloro-3,5-cyclohexanedione, 1,3-cyclohexanedione, 2-chloro-5,5-dimethyl, monochlorodimedon
PubChem CID 122278
IUPAC Name 2-chloro-5,5-dimethylcyclohexane-1,3-dione
SMILES CC1(CC(=O)C(C(=O)C1)Cl)C

Specifications

Melting Point 161°C to 164°C
Quantity 250 mg
Beilstein 2326749
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 174.62
Percent Purity 98%
Chemical Name or Material 2-Chloro-5,5-dimethyl-1,3-cyclohexanedione
compliance-icons
Product Identifier
  • 2-Chloro-5,5-dimethyl-1,3-cyclohexanedione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Chloro-5,5-dimethyl-1,3-cyclohexanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.