missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(1-chloro-1,3-dihydro-3-oxo-1-isobenzofuranyl)-benzenesulfonyl Chloride, TRC

Product Code. 30349096
Change view
Click to view available options
Quantity:
50 mg
500 mg
Product Code. Quantity
30349096 50 mg
30356957 500 mg
2 options
This item is not returnable. View return policy

Product Code. 30349096

Brand: Toronto Research Chemicals TRCC36435550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68592-11-0
Formula Weight 375.9131
InChI Formula InChI=1S/C14H7Cl3O4S/c15-11-6-5-8(7-12(11)22(17,19)20)14(16)10-4-2-1-3-9(10)13(18)21-14/h1-7H
IUPAC Name 2-chloro-5-(1-chloro-3-oxo-2-benzofuran-1-yl)benzenesulfonyl chloride
Molecular Formula C14H7Cl3O4S
Molecular Weight (g/mol) 377.63
SMILES O=C1OC(Cl)(C2=CC=C(Cl)C(=C2)S(=O)(=O)Cl)C=3C=CC=CC13
Synonym 3-Chloro-3-[4-chloro-3-(chlorosulfonyl)phenyl]-phthalide
Chemical Name or Material 2-Chloro-5-(1-chloro-1,3-dihydro-3-oxo-1-isobenzofuranyl)-benzenesulfonyl Chloride

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.