Learn More
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one, 97%
Description
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one is a reagent used in phosphorylation and phosphitylation of alcohols and in the formation of H-phosphonates. It is also reactive cyclic phosphitylating reagent involved in fast coupling rates and hydrolytic cleavage more readily compare to that of a acyclic analog.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one is a reagent used in phosphorylation and phosphitylation of alcohols and in the formation of H-phosphonates. It is also reactive cyclic phosphitylating reagent involved in fast coupling rates and hydrolytic cleavage more readily compare to that of a acyclic analog.
Solubility
Soluble in acetonitrile, dichloromethane, tetrahydrofuron and dimethylformamide.
Notes
Light and heat sensitive. Incompatible with water and strong oxidizing agents.
Specifications
Specifications
| Melting Point | 36°C to 40°C |
| Boiling Point | 127°C to 128°C (11 mmHg) |
| Flash Point | >110°C (230°F) |
| Quantity | 5 g |
| UN Number | UN3261 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in acetonitrile,dichloromethane,tetrahydrofuron and dimethylformamide. |
| Formula Weight | 202.54 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one |
Safety and Handling
- 2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: 2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.