missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one (90%), TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-4H-1,2,3-benzodioxaphosphorin-4-one |
| CAS | 5381-99-7 |
| Molecular Formula | C7 H4 Cl O3 P |
| Quantity | 100 g |
| InChI Formula | InChI=1S/C7H4ClO3P/c8-12-10-6-4-2-1-3-5(6)7(9)11-12/h1-4H |
| SMILES | ClP1OC(=O)c2ccccc2O1 |
| IUPAC Name | 2-chloro-1,3,2-benzodioxaphosphinin-4-one |
| Molecular Weight (g/mol) | 202.53 |
| Formula Weight | 201.9587 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction