missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)benzeneboronic acid, 96%

Product Code. 11324546
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324546 1 g 1g
11334546 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324546 Supplier Thermo Scientific Alfa Aesar Supplier No. L20103.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 254993-59-4 | C7H5BClF3O2 | 224.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 254993-59-4
Molecular Formula C7H5BClF3O2
Molecular Weight (g/mol) 224.37
MDL Number MFCD02684337
InChI Key JKSUCAFAUNSPLO-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl phenylboronic acid, 2-chloro-4-trifluoromethylphenylboronic acid, 2-chloro-4-trifluoromethyl benzeneboronic acid, 2-chloro-4-trifluoromethyl phenyl boronic acid, 2-chloro-4-trifluoromethyl phenylboronicacid, 2-chloro-4-trifluoromethylphenyl boronic acid, boronic acid, 2-chloro-4-trifluoromethyl phenyl, 2-fluoro-4-iodophenylboronicacid
PubChem CID 2783251
IUPAC Name [2-chloro-4-(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=C(Cl)C=C(C=C1)C(F)(F)F

Especificaciones

Melting Point 110°C to 112°C
Quantity 1 g
Beilstein 9263388
Formula Weight 224.38
Percent Purity 96%
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzeneboronic acid

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.