missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-4-(trifluoromethyl)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10250495
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10250495 25 g 25g
10754013 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10250495 Supplier Thermo Scientific Maybridge Supplier No. SEW01287EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-54-6
Molecular Formula C7H3Cl2F3O2S
Molecular Weight (g/mol) 279.05
MDL Number MFCD00052912
InChI Key NJXDBSSSDPOAFI-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl benzene-1-sulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulphonylchloride, 2-chloro-4-trifluoromethyl benzenesulphonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonylchloride, 2-chloro-4-trifluoromethyl benzenesulfonyl, 2-chloro-4-trifluoromethyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl, pubchem5104, acmc-1bwmt
PubChem CID 2736675
SMILES FC(F)(F)C1=CC(Cl)=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
IUPAC Name 2-chloro-4-(trifluoromethyl)benzenesulfonyl chloride
Formula Weight 279.06
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.