missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-picoline, 98%

Product Code. 10622623 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10622623 25 mL Glass bottle 25mL
10222722 5 mL Glass bottle 5mL
10581154 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10622623 Supplier Thermo Scientific Acros Supplier No. 368840250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3678-62-4 | C6H6ClN | 127.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3678-62-4
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00023418
InChI Key MZVSTDHRRYQFGI-UHFFFAOYSA-N
Synonym 2-chloro-4-picoline, pyridine, 2-chloro-4-methyl, 2-chloro-4-methyl-pyridine, 2-chloro-4-methyl pyridine, 2-chloro-gamma-picoline, 4-picoline, 2-chloro, zlchem 438, pubchem1192, acmc-209iop
PubChem CID 77248
IUPAC Name 2-chloro-4-methylpyridine
SMILES CC1=CC(=NC=C1)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1510g/mL
Boiling Point 194°C to 195°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5280 to 1.5300 (20°C, 589nm)
Quantity 25 mL
Beilstein 20,241
Specific Gravity 1.151
Formula Weight 127.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-picoline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-4-picoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.